Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DENND1A SIRNA Assays ›
Gene Symbol:
DENND1A
Gene Name:
DENN domain containing 1A
Gene Aliases:
FAM31A, KIAA1608
Chromosome Location:
Chr.9: 123379654 - 123930158 on Build GRCh38
UniGene ID:
Hs.744995
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
133564
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DENND1A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_020946.1
NP_065997.1
7, 8
690
NM_024820.2
NP_079096.2
7, 8
690
XM_005252109.2
XP_005252166.1
7, 8
539
XM_005252111.4
XP_005252168.1
7, 8
1506
XM_005252113.4
XP_005252170.1
7, 8
1495
XM_006717195.3
XP_006717258.1
7, 8
1507
XM_011518882.2
XP_011517184.1
7, 8
1509
XM_011518883.2
XP_011517185.1
7, 8
1510
XM_011518884.2
XP_011517186.1
6, 7
1460
XM_011518885.2
XP_011517187.1
6, 7
1202
XM_011518886.2
XP_011517188.1
7, 8
1507
XM_011518887.2
XP_011517189.1
7, 8
1498
XM_017014947.1
XP_016870436.1
6, 7
1465
XM_017014948.1
XP_016870437.1
7, 8
539
XM_017014949.1
XP_016870438.1
7, 8
1510
XM_017014950.1
XP_016870439.1
7, 8
1509
XM_017014951.1
XP_016870440.1
6, 7
483
XM_017014952.1
XP_016870441.1
6, 7
666
GenBank mRNA
AB046828.2
-
5, 6
554
AK024782.1
-
7, 8
678
AK074151.1
-
2, 3
118
AK225104.1
-
7, 8
678
AK295710.1
-
5, 6
551
BC009616.2
-
6, 7
658
BC028061.1
-
6, 7
480
BC039703.1
-
7, 8
543
4999484
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
protein transport
endocytic recycling
regulation of Rab protein signal transduction
positive regulation of GTPase activity
synaptic vesicle endocytosis
Rab guanyl-nucleotide exchange factor activity
SH3 domain binding
phosphatidylinositol-3-phosphate binding
phosphatidylinositol phosphate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7745c474b-b2rzg:80/100.66.78.226:80.
git-commit: c3c9db9cc82507548abdd0ae6a4ade3f81f66292
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.20.0-2024.11.45-1.0