Search Thermo Fisher Scientific
Gene Symbol: TAF7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168474.1 | NP_001161946.1 | 13 | 1787 | ||
NM_024885.3 | NP_079161.3 | 13 | 1955 | |||
XM_005262152.1 | XP_005262209.1 | 13 | 1790 | |||
XM_006724664.1 | XP_006724727.1 | 13 | 1957 | |||
GenBank mRNA | AK026810.1 | - | 11 | 1711 | ||
BC043391.1 | - | 13 | 1941 | 6154641 |