Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S1PR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166215.1 | NP_001159687.1 | 2 | 1962 | ||
NM_030760.4 | NP_110387.1 | 2 | 1846 | |||
GenBank mRNA | AF088014.1 | - | 1 | 310 | ||
AF331840.1 | - | 1 | 1784 | |||
AF331841.1 | - | 2 | 1353 | |||
AF331842.1 | - | 2 | 900 | |||
AK074661.1 | - | 2 | 1898 | |||
AK094880.1 | - | 3 | 1746 | |||
BC034703.1 | - | 2 | 1829 | 4754850 | ||
BC067781.1 | - | 3 | 1752 | 5286249 |
Set Membership: |
![]() |