Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003811.1 | NP_001003811.1 | 9 | 768 | ||
NM_031276.2 | NP_112566.2 | 8 | 646 | |||
XM_011530994.1 | XP_011529296.1 | 8 | 640 | |||
XM_017029649.1 | XP_016885138.1 | 8 | 640 | |||
GenBank mRNA | AF285594.1 | - | 7 | 551 | ||
AK057523.1 | - | 9 | 762 | |||
AK292471.1 | - | 8 | 640 | |||
BC036016.1 | - | 8 | 646 | 5270970 |