Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IFT122 SIRNA Assays ›
Gene Symbol:
IFT122
Gene Name:
intraflagellar transport 122
Gene Aliases:
CED, CED1, SPG, WDR10, WDR10p, WDR140
Chromosome Location:
Chr.3: 129440036 - 129520507 on Build GRCh38
UniGene ID:
Hs.655284
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
133661
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IFT122
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001280541.1
NP_001267470.1
10
1153
NM_001280545.1
NP_001267474.1
11
1328
NM_001280546.1
NP_001267475.1
8
921
NM_018262.3
NP_060732.2
9
1000
NM_052985.3
NP_443711.2
11
1330
NM_052989.2
NP_443715.1
10
1177
NM_052990.2
NP_443716.1
7
844
XM_005247609.1
XP_005247666.1
9
965
XM_006713689.1
XP_006713752.1
10
1142
XM_006713691.2
XP_006713754.1
10
1142
XM_006713692.2
XP_006713755.1
9
965
XM_006713695.2
XP_006713758.1
10
1142
XM_011512972.2
XP_011511274.1
10
1142
XM_017006817.1
XP_016862306.1
10
1142
XM_017006818.1
XP_016862307.1
10
1142
XM_017006819.1
XP_016862308.1
10
1142
XM_017006820.1
XP_016862309.1
10
1142
XM_017006821.1
XP_016862310.1
10
1142
XM_017006822.1
XP_016862311.1
8
933
XM_017006823.1
XP_016862312.1
9
965
XM_017006824.1
XP_016862313.1
9
965
XM_017006825.1
XP_016862314.1
9
965
XM_017006826.1
XP_016862315.1
9
965
XM_017006827.1
XP_016862316.1
9
965
XM_017006828.1
XP_016862317.1
7
756
XM_017006829.1
XP_016862318.1
7
756
XM_017006830.1
XP_016862319.1
11
1140
XM_017006831.1
XP_016862320.1
9
1061
XM_017006832.1
XP_016862321.1
10
1040
XM_017006833.1
XP_016862322.1
7
756
XM_017006834.1
XP_016862323.1
7
756
XM_017006835.1
XP_016862324.1
8
884
XM_017006836.1
XP_016862325.1
10
963
XM_017006837.1
XP_016862326.1
1
67
XM_017006838.1
XP_016862327.1
6
507
GenBank mRNA
AF244930.1
-
10
1074
AF244931.1
-
11
1227
AF302154.1
-
10
959
AK001759.1
-
9
813
AK022525.1
-
9
911
AK023000.1
-
9
642
AK024435.1
-
8
688
AK122965.1
-
4
795
AK124140.1
-
11
1131
AK223095.1
-
9
812
AK293852.1
-
7
658
AK294515.1
-
5
549
AK298526.1
-
6
421
AK299903.1
-
8
634
AK315103.1
-
10
977
AL133565.1
-
8
754
BC003045.1
-
7
636
2823536
BC004238.2
-
7
636
2823536
BC028353.2
-
10
917
4836844
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
signal transduction downstream of smoothened
embryonic body morphogenesis
negative regulation of smoothened signaling pathway involved in ventral spinal cord patterning
embryonic heart tube development
embryonic forelimb morphogenesis
intraciliary anterograde transport
intraciliary retrograde transport
intraciliary transport involved in cilium morphogenesis
cilium assembly
embryonic digit morphogenesis
negative regulation of smoothened signaling pathway
camera-type eye morphogenesis
negative regulation of epithelial cell proliferation
limb development
cilium morphogenesis
ciliary receptor clustering involved in smoothened signaling pathway
smoothened signaling pathway involved in dorsal/ventral neural tube patterning
embryonic heart tube left/right pattern formation
protein localization to cilium
establishment of protein localization to organelle
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-vrvhj:80/10.239.161.228:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0