Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IFT122 SIRNA Assays ›
Gene Symbol:
IFT122
Gene Name:
intraflagellar transport 122
Gene Aliases:
CED, CED1, SPG, WDR10, WDR10p, WDR140
Chromosome Location:
Chr.3: 129440036 - 129520507 on Build GRCh38
UniGene ID:
Hs.655284
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
133663
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IFT122
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001280541.1
NP_001267470.1
18, 19
2487
NM_001280545.1
NP_001267474.1
19, 20
2662
NM_001280546.1
NP_001267475.1
16, 17
2255
NM_018262.3
NP_060732.2
17, 18
2334
NM_052985.3
NP_443711.2
19, 20
2664
NM_052989.2
NP_443715.1
18, 19
2511
NM_052990.2
NP_443716.1
15, 16
2178
XM_005247609.1
XP_005247666.1
17, 18
2299
XM_006713689.1
XP_006713752.1
18, 19
2476
XM_006713691.2
XP_006713754.1
17, 18
2422
XM_006713692.2
XP_006713755.1
16, 17
2245
XM_006713695.2
XP_006713758.1
18, 19
2476
XM_011512972.2
XP_011511274.1
18, 19
2476
XM_017006817.1
XP_016862306.1
18, 19
2476
XM_017006818.1
XP_016862307.1
18, 19
2476
XM_017006819.1
XP_016862308.1
17, 18
2422
XM_017006820.1
XP_016862309.1
17, 18
2422
XM_017006821.1
XP_016862310.1
17, 18
2422
XM_017006822.1
XP_016862311.1
16, 17
2267
XM_017006823.1
XP_016862312.1
17, 18
2299
XM_017006824.1
XP_016862313.1
17, 18
2299
XM_017006825.1
XP_016862314.1
16, 17
2245
XM_017006826.1
XP_016862315.1
16, 17
2245
XM_017006827.1
XP_016862316.1
16, 17
2245
XM_017006828.1
XP_016862317.1
15, 16
2090
XM_017006829.1
XP_016862318.1
15, 16
2090
XM_017006830.1
XP_016862319.1
19, 20
2474
XM_017006831.1
XP_016862320.1
17, 18
2395
XM_017006832.1
XP_016862321.1
18, 19
2374
XM_017006833.1
XP_016862322.1
14, 15
2036
XM_017006834.1
XP_016862323.1
14, 15
2036
XM_017006835.1
XP_016862324.1
16, 17
2218
XM_017006836.1
XP_016862325.1
17, 18
2243
XM_017006837.1
XP_016862326.1
9, 10
1420
XM_017006838.1
XP_016862327.1
14, 15
1860
GenBank mRNA
AF244930.1
-
18, 19
2408
AF244931.1
-
19, 20
2561
AF302154.1
-
18, 19
2293
AK001759.1
-
17, 18
2147
AK001955.1
-
7, 8
956
AK022525.1
-
17, 18
2245
AK023000.1
-
17, 18
1976
AK024435.1
-
16, 17
2022
AK094922.1
-
2, 3
377
AK122965.1
-
11, 12
2012
AK124140.1
-
19, 20
2465
AK223095.1
-
17, 18
2146
AK293852.1
-
14, 15
1938
AK298526.1
-
14, 15
1755
AK299903.1
-
16, 17
1968
AK307463.1
-
5, 6
741
AK315103.1
-
18, 19
2311
AL133565.1
-
16, 17
2088
AL833851.1
-
7, 8
958
BC003045.1
-
15, 16
1970
2823536
BC004238.2
-
15, 16
1970
2823536
BC028353.2
-
18, 19
2251
4836844
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_001057925, XM_006237067, BC087667.
This siRNA also targets the following transcripts in Mouse: NM_001167763, NM_031177, AK167412, AK167330, AK044456, and others.
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
signal transduction downstream of smoothened
embryonic body morphogenesis
negative regulation of smoothened signaling pathway involved in ventral spinal cord patterning
embryonic heart tube development
embryonic forelimb morphogenesis
intraciliary anterograde transport
intraciliary retrograde transport
intraciliary transport involved in cilium morphogenesis
cilium assembly
embryonic digit morphogenesis
negative regulation of smoothened signaling pathway
camera-type eye morphogenesis
negative regulation of epithelial cell proliferation
limb development
cilium morphogenesis
ciliary receptor clustering involved in smoothened signaling pathway
smoothened signaling pathway involved in dorsal/ventral neural tube patterning
embryonic heart tube left/right pattern formation
protein localization to cilium
establishment of protein localization to organelle
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-8tvtn:80/10.239.165.222:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0