Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198532.1 | NP_001185461.1 | 6 | 693 | ||
NM_001198533.1 | NP_001185462.1 | 7 | 919 | |||
NM_018002.3 | NP_060472.2 | 7 | 919 | |||
NM_181354.4 | NP_851999.2 | 5 | 749 | |||
XM_006716595.2 | XP_006716658.1 | 6 | 688 | |||
XM_017013589.1 | XP_016869078.1 | 5 | 718 | |||
XM_017013590.1 | XP_016869079.1 | 5 | 559 | |||
XM_017013591.1 | XP_016869080.1 | 5 | 649 | |||
XM_017013592.1 | XP_016869081.1 | 5 | 396 | |||
XM_017013593.1 | XP_016869082.1 | 5 | 623 | |||
XM_017013594.1 | XP_016869083.1 | 5 | 623 | |||
XM_017013595.1 | XP_016869084.1 | 5 | 718 | |||
XM_017013596.1 | XP_016869085.1 | 7 | 891 | |||
XM_017013597.1 | XP_016869086.1 | 5 | 552 | |||
XM_017013598.1 | XP_016869087.1 | 5 | 623 | |||
XM_017013599.1 | XP_016869088.1 | 5 | 718 | |||
XM_017013600.1 | XP_016869089.1 | 5 | 575 | |||
GenBank mRNA | AK096148.1 | - | 5 | 429 | ||
AK124441.1 | - | 7 | 852 | |||
AK292899.1 | - | 5 | 701 | |||
AK296561.1 | - | 6 | 693 | |||
AL833193.1 | - | 4 | 330 | |||
BC019691.1 | - | 5 | 566 | |||
BC032710.2 | - | 5 | 691 | 5496492 | ||
BC035484.1 | - | 5 | 703 | |||
FN650108.1 | - | 6 | 729 |