Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198532.1 | NP_001185461.1 | 7 | 878 | ||
NM_001198533.1 | NP_001185462.1 | 8 | 1104 | |||
NM_018002.3 | NP_060472.2 | 8 | 1104 | |||
NM_181354.4 | NP_851999.2 | 6 | 934 | |||
XM_006716595.2 | XP_006716658.1 | 7 | 873 | |||
XM_017013589.1 | XP_016869078.1 | 6 | 903 | |||
XM_017013590.1 | XP_016869079.1 | 6 | 744 | |||
XM_017013591.1 | XP_016869080.1 | 6 | 834 | |||
XM_017013592.1 | XP_016869081.1 | 6 | 581 | |||
XM_017013593.1 | XP_016869082.1 | 6 | 808 | |||
XM_017013594.1 | XP_016869083.1 | 6 | 808 | |||
GenBank mRNA | AK096148.1 | - | 6 | 614 | ||
AK124441.1 | - | 8 | 1037 | |||
AK292899.1 | - | 6 | 886 | |||
AK296561.1 | - | 7 | 878 | |||
AL833193.1 | - | 5 | 515 | |||
BC019691.1 | - | 6 | 751 | |||
BC032710.2 | - | 6 | 876 | 5496492 | ||
BC035484.1 | - | 6 | 888 | |||
FN650108.1 | - | 7 | 914 |