Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198532.1 | NP_001185461.1 | 7, 8 | 956 | ||
NM_001198533.1 | NP_001185462.1 | 8, 9 | 1182 | |||
NM_018002.3 | NP_060472.2 | 8, 9 | 1182 | |||
NM_181354.4 | NP_851999.2 | 6, 7 | 1012 | |||
XM_006716595.2 | XP_006716658.1 | 7, 8 | 951 | |||
XM_017013589.1 | XP_016869078.1 | 6, 7 | 981 | |||
XM_017013590.1 | XP_016869079.1 | 6, 7 | 822 | |||
XM_017013591.1 | XP_016869080.1 | 6, 7 | 912 | |||
XM_017013592.1 | XP_016869081.1 | 6, 7 | 659 | |||
XM_017013593.1 | XP_016869082.1 | 6, 7 | 886 | |||
XM_017013594.1 | XP_016869083.1 | 6, 7 | 886 | |||
GenBank mRNA | AK096148.1 | - | 6, 7 | 692 | ||
AK124441.1 | - | 8, 9 | 1115 | |||
AK292899.1 | - | 6, 7 | 964 | |||
AK296561.1 | - | 7, 8 | 956 | |||
AL833193.1 | - | 5, 6 | 593 | |||
BC019691.1 | - | 6, 7 | 829 | |||
BC032710.2 | - | 6, 7 | 954 | 5496492 | ||
BC035484.1 | - | 6, 7 | 966 | |||
FN650108.1 | - | 7, 8 | 992 |