Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018518.4 | NP_060988.3 | 8, 9 | 1234 | ||
NM_182751.2 | NP_877428.1 | 8, 9 | 1237 | |||
XM_011519538.2 | XP_011517840.1 | 8, 9 | 1207 | |||
GenBank mRNA | AB042719.1 | - | 8, 9 | 1201 | ||
AK055695.1 | - | 8, 9 | 1210 | |||
AK292701.1 | - | 8, 9 | 1212 | |||
AL136840.1 | - | 7, 8 | 1200 | |||
BC004876.2 | - | 1, 2 | 152 | |||
BC009108.1 | - | 3, 4 | 452 | |||
BC101727.1 | - | 8, 9 | 1206 | 8069233 | ||
BC143490.1 | - | 8, 9 | 1199 | 9051998 |