Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: BMP2K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017593.3 | NP_060063.2 | 2 | 411 | ||
NM_198892.1 | NP_942595.1 | 2 | 411 | |||
XM_005263117.1 | XP_005263174.1 | 2 | 386 | |||
XM_011532102.1 | XP_011530404.1 | 2 | 386 | |||
XM_017008381.1 | XP_016863870.1 | 2 | 163 | |||
GenBank mRNA | AK292147.1 | - | 2 | 387 | ||
AL137661.1 | - | 2 | 447 | |||
BC036021.1 | - | 2 | 372 | 5272264 |