Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005813.4 | NP_005804.1 | 3, 4 | 1122 | ||
XM_005264237.3 | XP_005264294.1 | 4, 5 | 1395 | |||
XM_005264238.3 | XP_005264295.1 | 3, 4 | 371 | |||
XM_017003741.1 | XP_016859230.1 | 5, 6 | 1400 | |||
XM_017003742.1 | XP_016859231.1 | 4, 5 | 439 | |||
GenBank mRNA | AB015982.2 | - | 3, 4 | 1122 | ||
BC012820.2 | - | 3, 4 | 1081 | |||
BC030706.2 | - | 3, 4 | 1067 | 5266769 |