Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITSN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006277.2 | NP_006268.2 | 3 | 333 | ||
NM_019595.3 | NP_062541.3 | 3 | 333 | |||
NM_147152.2 | NP_671494.2 | 3 | 333 | |||
GenBank mRNA | AB033082.1 | - | 3 | 240 | ||
AF182198.1 | - | 3 | 312 | |||
AF182199.1 | - | 3 | 312 | |||
AF248540.1 | - | 2 | 114 | |||
BC020921.2 | - | 2 | 110 | |||
BC038963.1 | - | 3 | 324 | |||
BC146779.1 | - | 3 | 240 | 9007231 | ||
DA382952.1 | - | 3 | 583 |