Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004720.2 | NP_001004720.1 | 4 | 1156 | ||
NM_001004722.3 | NP_001004722.1 | 4 | 544 | |||
NM_003581.4 | NP_003572.2 | 5 | 1266 | |||
XM_006712797.3 | XP_006712860.1 | 5 | 1922 | |||
XM_011511991.2 | XP_011510293.1 | 5 | 1360 | |||
XM_011511992.2 | XP_011510294.2 | 3 | 488 | |||
XM_017005103.1 | XP_016860592.1 | 6 | 1524 | |||
XM_017005104.1 | XP_016860593.1 | 5 | 1536 | |||
XM_017005105.1 | XP_016860594.1 | 5 | 1273 | |||
GenBank mRNA | AF043119.1 | - | 3 | 1018 | ||
AF047487.1 | - | 4 | 1122 | |||
BC000103.2 | - | 4 | 1040 | 3505427 | ||
BC007195.1 | - | 4 | 1047 | 3608149 | ||
BG480643.1 | - | 4 | 546 | |||
KU178418.1 | - | 3 | 990 | |||
KU178419.1 | - | 2 | 268 |