Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142522.1 | NP_001135994.1 | 2 | 439 | ||
NM_012177.3 | NP_036309.1 | 2 | 279 | |||
GenBank mRNA | AF129535.1 | - | 2 | 231 | ||
AJ420499.1 | - | 2 | 299 | |||
AK026197.1 | - | 3 | 188 | |||
AK055221.1 | - | 2 | 413 | |||
AK314456.1 | - | 2 | 261 | |||
AY079515.1 | - | 2 | 170 | |||
BC018905.1 | - | 2 | 221 | 3960052 | ||
BG944882.1 | - | 2 | 251 | |||
BI838832.1 | - | 2 | 470 | |||
DB122061.1 | - | 2 | 279 |