Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003630.2 | NP_003621.1 | 3, 4 | 557 | ||
GenBank mRNA | AB035307.1 | - | 3, 4 | 441 | ||
AJ001625.1 | - | 3, 4 | 358 | |||
AJ131389.1 | - | 3, 4 | 354 | |||
AK025644.1 | - | 423 | ||||
AK300907.1 | - | 3, 4 | 423 | |||
AY277600.1 | - | 3, 4 | 314 | |||
BC014551.1 | - | 3, 4 | 495 | 4333011 | ||
BC015506.1 | - | 3, 4 | 456 | 3914956 | ||
BP377713.1 | - | 3, 4 | 557 | |||
CR542062.1 | - | 3, 4 | 295 |