Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114094.1 | NP_001107566.1 | 16 | 1705 | ||
NM_001258440.1 | NP_001245369.1 | 16 | 1558 | |||
NM_001258441.1 | NP_001245370.1 | 15 | 1489 | |||
NM_001258442.1 | NP_001245371.1 | 13 | 1243 | |||
NM_013314.3 | NP_037446.1 | 17 | 1774 | |||
NR_047680.1 | 16 | 1538 | ||||
NR_047681.1 | 15 | 1486 | ||||
NR_047682.1 | 15 | 1481 | ||||
NR_047683.1 | 14 | 1419 | ||||
XM_011539728.2 | XP_011538030.1 | 17 | 1651 | |||
XM_017016159.1 | XP_016871648.1 | 16 | 1583 | |||
GenBank mRNA | AF068180.1 | - | 17 | 1749 | ||
AF068181.1 | - | 16 | 1680 | |||
AK027779.1 | - | 17 | 1709 | |||
AK225546.1 | - | 16 | 1689 | |||
BC018906.2 | - | 17 | 1688 | 4053894 | ||
BC110298.1 | - | 16 | 1530 |