Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013373.3 | NP_037505.1 | 11 | 3363 | ||
GenBank mRNA | AB033118.1 | - | 7 | 2747 | ||
AY894890.1 | - | 11 | 3362 | |||
BC009442.1 | - | 1 | 861 | |||
BC053544.1 | - | 11 | 3362 | 5730191 |