Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014447.2 | NP_001014447.1 | 7 | 1373 | ||
NM_001014448.2 | NP_001014448.1 | 7 | 1636 | |||
NM_003652.3 | NP_003643.2 | 6 | 1340 | |||
GenBank mRNA | AK056317.1 | - | 7 | 1372 | ||
AK127844.1 | - | 4 | 2709 | |||
BC006393.2 | - | 7 | 1540 | |||
BC018913.2 | - | 7 | 1540 | |||
BC080539.1 | - | 7 | 1307 | 5020194 | ||
U83411.1 | - | 6 | 1205 |