Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013446.3 | NP_038474.2 | 8 | 2789 | ||
NR_117084.1 | 6 | 2332 | ||||
XM_011515996.1 | XP_011514298.1 | 7 | 2578 | |||
XM_011515997.2 | XP_011514299.1 | 8 | 2846 | |||
XM_011515998.1 | XP_011514300.1 | 8 | 2531 | |||
GenBank mRNA | AB209298.1 | - | 7 | 3112 | ||
AF192784.1 | - | 8 | 2684 | |||
AK024987.1 | - | 3 | 1534 | |||
AK027137.1 | - | 7 | 2301 | |||
AK127030.1 | - | 9 | 3406 | |||
AL136812.1 | - | 8 | 2687 | |||
BC025955.1 | - | 8 | 2722 | 4869484 | ||
BC037400.1 | - | 8 | 2574 | 5176176 | ||
BC064838.1 | - | 8 | 2568 | 6173715 |