Search Thermo Fisher Scientific
Gene Symbol: ADAM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288973.1 | NP_001275902.1 | 8 | 1103 | ||
NM_001288974.1 | NP_001275903.1 | 8 | 1103 | |||
NM_001288975.1 | NP_001275904.1 | 8 | 1103 | |||
NM_003474.5 | NP_003465.3 | 8 | 1112 | |||
NM_021641.4 | NP_067673.2 | 8 | 1112 | |||
XM_017016705.1 | XP_016872194.1 | 7 | 1015 | |||
GenBank mRNA | AF023476.2 | - | 8 | 1005 | ||
AF023477.2 | - | 8 | 1005 | |||
AK127856.1 | - | 8 | 1008 | |||
AK291629.1 | - | 8 | 922 | |||
AY358878.1 | - | 8 | 1000 | |||
BC060804.1 | - | 8 | 1042 | 30348514 | ||
KF444157.1 | - | 8 | 1103 |