Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304476.1 | NP_001291405.1 | 7 | 1664 | ||
NM_001304477.1 | NP_001291406.1 | 6 | 1169 | |||
NM_014178.7 | NP_054897.4 | 6 | 1811 | |||
XM_006720121.3 | XP_006720184.1 | 6 | 1174 | |||
XM_011536669.2 | XP_011534971.1 | 5 | 1206 | |||
XM_017021231.1 | XP_016876720.1 | 6 | 1543 | |||
XM_017021232.1 | XP_016876721.1 | 7 | 4139 | |||
XM_017021233.1 | XP_016876722.1 | 7 | 1491 | |||
XM_017021234.1 | XP_016876723.1 | 7 | 1264 | |||
XM_017021235.1 | XP_016876724.1 | 6 | 4052 | |||
XM_017021236.1 | XP_016876725.1 | 7 | 1468 | |||
XM_017021237.1 | XP_016876726.1 | 7 | 1297 | |||
XM_017021238.1 | XP_016876727.1 | 6 | 1235 | |||
XM_017021239.1 | XP_016876728.1 | 6 | 1233 | |||
XM_017021240.1 | XP_016876729.1 | 6 | 1155 | |||
XM_017021241.1 | XP_016876730.1 | 6 | 1772 | |||
XM_017021242.1 | XP_016876731.1 | 7 | 1237 | |||
GenBank mRNA | AL834346.1 | - | 6 | 1149 | ||
BC067278.1 | - | 6 | 1811 | 6178786 |