Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SACS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278055.1 | NP_001264984.1 | 5 | 635 | ||
NM_014363.5 | NP_055178.3 | 7 | 1166 | |||
XM_005266338.2 | XP_005266395.1 | 7 | 1166 | |||
XM_011535039.2 | XP_011533341.1 | 6 | 774 | |||
XM_017020539.1 | XP_016876028.1 | 6 | 774 | |||
GenBank mRNA | AK125458.1 | - | 5 | 635 | ||
AW295072.1 | - | 4 | 390 | |||
BX640926.1 | - | 3 | 249 | |||
CR987846.1 | - | 4 | 383 |