Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170760.1 | NP_001164231.1 | 8, 9 | 870 | ||
NM_001170761.1 | NP_001164232.1 | 8, 9 | 870 | |||
NM_014370.3 | NP_055185.2 | 9 | 873 | |||
GenBank mRNA | AF027406.1 | - | 8, 9 | 808 | ||
AK301749.1 | - | 9 | 833 | |||
AK314828.1 | - | 9 | 833 | |||
BC092416.1 | - | 9 | 819 | 30529915 | ||
BC117124.1 | - | 8, 9 | 864 | 40125675 | ||
DQ099381.1 | - | 8, 9 | 784 | |||
U82808.1 | - | 8, 9 | 688 |