Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170760.1 | NP_001164231.1 | 13, 14 | 1519 | ||
NM_001170761.1 | NP_001164232.1 | 14, 15 | 1420 | |||
NM_014370.3 | NP_055185.2 | 13, 14 | 1522 | |||
GenBank mRNA | AF027406.1 | - | 14, 15 | 1358 | ||
AK301749.1 | - | 13, 14 | 1482 | |||
AK314828.1 | - | 13, 14 | 1482 | |||
BC092416.1 | - | 13, 14 | 1468 | 30529915 | ||
BC117124.1 | - | 13, 14 | 1513 | 40125675 | ||
DQ099381.1 | - | 13, 14 | 1433 | |||
U82808.1 | - | 14, 15 | 1238 |