Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258448.1 | NP_001245377.1 | 9 | 1378 | ||
NM_001300921.1 | NP_001287850.1 | 9 | 1444 | |||
NM_014386.3 | NP_055201.2 | 9 | 1444 | |||
XM_011543318.2 | XP_011541620.1 | 8 | 1358 | |||
XM_011543321.1 | XP_011541623.1 | 9 | 1378 | |||
XM_017009343.1 | XP_016864832.1 | 9 | 1460 | |||
XM_017009344.1 | XP_016864833.1 | 8 | 1568 | |||
XM_017009345.1 | XP_016864834.1 | 8 | 1238 | |||
GenBank mRNA | AF118125.1 | - | 9 | 1411 | ||
AF182034.1 | - | 9 | 1414 | |||
AK301924.1 | - | 9 | 1347 |