Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014441.2 | NP_055256.1 | 7 | 1598 | ||
GenBank mRNA | AB026265.1 | - | 7 | 1549 | ||
AF227924.1 | - | 7 | 1570 | |||
AY358913.1 | - | 7 | 1545 | |||
BC035365.1 | - | 7 | 1552 | 5176457 |