Gene Symbol: PCDH17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040429.2 | NP_001035519.1 | 1 | 1512 | ||
XM_005266357.2 | XP_005266414.1 | 2 | 2957 | |||
XM_005266358.2 | XP_005266415.1 | 1 | 3063 | |||
XM_017020547.1 | XP_016876036.1 | 1 | 3063 | |||
GenBank mRNA | AF029343.1 | - | 1 | 1357 | ||
AK289980.1 | - | 1 | 1355 | |||
BC028165.1 | - | 1 | 646 | 5245587 |