- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: PCDH17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040429.2 | NP_001035519.1 | 4 | 4802 | ||
XM_005266357.2 | XP_005266414.1 | 5 | 6247 |