Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014584.2 | NP_055399.1 | 16 | 3341 | ||
XM_011536684.2 | XP_011534986.1 | 15 | 3148 | |||
GenBank mRNA | AF081886.1 | - | 16 | 3117 |