Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L13 SIRNA Assays ›
Gene Symbol:
BCL2L13
Gene Name:
BCL2 like 13
Gene Aliases:
BCL-RAMBO, Bcl2-L-13, MIL1
Chromosome Location:
Chr.22: 17628855 - 17730855 on Build GRCh38
UniGene ID:
Hs.631672
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
134468
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270726.1
NP_001257655.1
6
820
NM_001270727.1
NP_001257656.1
5
676
NM_001270728.1
NP_001257657.1
4
656
NM_001270729.1
NP_001257658.1
6
834
NM_001270730.1
NP_001257659.1
5
663
NM_001270731.1
NP_001257660.1
6
921
NM_001270732.1
NP_001257661.1
5
764
NM_001270733.1
NP_001257662.1
2
206
NM_001270734.1
NP_001257663.1
5
777
NM_001270735.1
NP_001257664.1
4
620
NM_015367.3
NP_056182.2
7
991
NR_073068.1
5
589
NR_073069.1
5
572
XM_011546119.1
XP_011544421.1
7
1354
XM_011546120.1
XP_011544422.1
7
1356
XM_011546121.2
XP_011544423.1
8
1339
XM_011546123.1
XP_011544425.1
6
719
XM_017028725.1
XP_016884214.1
6
1210
XM_017028726.1
XP_016884215.1
9
1640
XM_017028727.1
XP_016884216.1
7
789
XM_017028728.1
XP_016884217.1
7
1368
XM_017028729.1
XP_016884218.1
7
863
XM_017028730.1
XP_016884219.1
5
577
XM_017028731.1
XP_016884220.1
6
1197
XM_017028732.1
XP_016884221.1
4
440
XM_017028734.1
XP_016884223.1
6
1298
GenBank mRNA
AF146568.1
-
4
412
AF183411.1
-
573
AF325209.1
-
6
638
AF539453.1
-
5
522
AK056248.1
-
6
677
AK291449.1
-
5
584
AK294286.1
-
4
483
AK301394.1
-
2
120
AK314489.1
-
7
818
AL133029.1
-
2
93
AL831982.2
-
5
551
BC003032.1
-
3
251
BC007658.2
-
7
750
3343994
BX354578.2
-
6
700
BX407004.2
-
6
573
CN420914.1
-
5
571
CR456521.1
-
7
894
DA154984.1
-
5
490
DA169152.1
-
4
469
DA478149.1
-
5
506
DC341613.1
-
2
210
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
protein binding
cysteine-type endopeptidase activator activity involved in apoptotic process
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0