Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GORASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201428.1 | NP_001188357.1 | 6 | 744 | ||
NM_015530.4 | NP_056345.3 | 6 | 1499 | |||
XM_006712408.2 | XP_006712471.1 | 6 | 822 | |||
GenBank mRNA | AK023082.1 | - | 6 | 771 | ||
AK023201.1 | - | 6 | 784 | |||
AK027202.1 | - | 1 | 951 | |||
AK027349.1 | - | 6 | 698 | |||
AK125761.1 | - | 6 | 1499 | |||
AK225810.1 | - | 6 | 781 | |||
AK293640.1 | - | 5 | 631 | |||
AK296698.1 | - | 7 | 808 | |||
AK298020.1 | - | 6 | 778 | |||
AK298771.1 | - | 4 | 365 | |||
AK315508.1 | - | 6 | 800 | |||
AL117430.1 | - | 6 | 710 | |||
BC007770.2 | - | 6 | 733 | 4298251 |