Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031623.2 | NP_001026794.1 | 3 | 341 | ||
NM_001257273.1 | NP_001244202.1 | 3 | 361 | |||
NM_015555.2 | NP_056370.2 | 3 | 341 | |||
XM_005248994.1 | XP_005249051.1 | 3 | 361 | |||
XM_011514460.1 | XP_011512762.1 | 2 | 100 | |||
XM_011514462.2 | XP_011512764.1 | 3 | 361 | |||
XM_011514463.2 | XP_011512765.1 | 2 | 118 | |||
GenBank mRNA | AB011148.1 | - | 3 | 160 | ||
AK225806.1 | - | 3 | 168 | |||
AK302577.1 | - | 3 | 206 | |||
AY055204.1 | - | 3 | 342 | |||
BC021712.2 | - | 3 | 258 | 4821000 | ||
BC042450.1 | - | 1 | 27 | |||
BC058853.1 | - | 1 | 27 | |||
BC098382.1 | - | 3 | 184 | 5295845 | ||
BX648081.1 | - | 3 | 187 |