Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320766.1 | NP_001307695.1 | 2 | 294 | ||
NM_015565.2 | NP_056380.2 | 2 | 291 | |||
XM_006723987.3 | XP_006724050.1 | 2 | 289 | |||
XM_017028316.1 | XP_016883805.1 | 2 | 152 | |||
GenBank mRNA | AB018257.2 | - | 2 | 182 | ||
AB621822.1 | - | 2 | 198 | |||
AF161350.1 | - | 2 | 179 | |||
AK001915.1 | - | 2 | 238 | |||
BC029371.1 | - | 2 | 198 | |||
BC140790.1 | - | 2 | 204 | 9021658 | ||
BC150284.1 | - | 2 | 182 | 8860138 | ||
DB452437.1 | - | 2 | 294 | |||
HY001556.1 | - | 2 | 299 |