Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLTSCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015710.4 | NP_056525.2 | 6 | 765 | ||
GenBank mRNA | AF182076.1 | - | 6 | 748 | ||
AF296124.1 | - | 6 | 711 | |||
AK024486.1 | - | 6 | 736 | |||
AK225915.1 | - | 6 | 794 | |||
AK297877.1 | - | 6 | 751 | |||
AK303918.1 | - | 752 | ||||
AK309788.1 | - | 6 | 753 | |||
AL122063.1 | - | 2 | 92 | |||
AL359335.1 | - | 6 | 684 | |||
AL359336.1 | - | 6 | 708 | |||
AY535000.1 | - | 6 | 741 | |||
BC004229.2 | - | 6 | 725 | |||
BC006311.2 | - | 6 | 747 | 4111480 | ||
BC007248.1 | - | 6 | 729 | |||
BC010095.2 | - | 6 | 784 | 3501649 | ||
BC013203.1 | - | 6 | 753 | |||
BC013235.1 | - | 6 | 736 | |||
BC013307.1 | - | 6 | 738 | |||
BC014009.2 | - | 6 | 735 | |||
BC039869.1 | - | 6 | 737 | |||
CX866974.1 | - | 6 | 765 |