Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276310.1 | NP_001263239.1 | 5 | 765 | ||
NM_015894.3 | NP_056978.2 | 5 | 855 | |||
NR_075070.1 | 5 | 952 | ||||
GenBank mRNA | AK074860.1 | - | 5 | 666 | ||
AK131326.1 | - | 1 | 531 | |||
AK298903.1 | - | 5 | 765 | |||
AK310010.1 | - | 5 | 787 | |||
BC009381.2 | - | 5 | 712 | 4124704 | ||
BC025234.1 | - | 5 | 712 | 4894236 | ||
BC047671.1 | - | 5 | 602 | |||
BX449528.1 | - | 2 | 154 |