Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015918.3 | NP_057002.2 | 5 | 732 | ||
NM_198202.1 | NP_937845.1 | 4 | 586 | |||
XM_011538441.1 | XP_011536743.1 | 5 | 844 | |||
GenBank mRNA | AF070660.1 | - | 5 | 716 | ||
AF117232.1 | - | 5 | 716 | |||
AJ306296.1 | - | 5 | 719 | |||
AK223206.1 | - | 5 | 704 | |||
AK303144.1 | - | 4 | 567 | |||
BC012505.1 | - | 5 | 719 | 4708484 | ||
BU155588.1 | - | 575 |