Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETDB2-PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320727.1 | NP_001307656.1 | 20 | 3592 | ||
NR_135324.1 | 21 | 3796 |
Gene Symbol: PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040443.2 | NP_001035533.1 | 10 | 1596 | ||
NM_001040444.2 | NP_001035534.1 | 10 | 1272 | |||
NR_135322.1 | 9 | 1474 | ||||
NR_135323.1 | 12 | 1573 | ||||
GenBank mRNA | AB011031.1 | - | 10 | 1245 | ||
AF155105.1 | - | 10 | 1217 | |||
AK026228.1 | - | 10 | 1236 | |||
AK225986.1 | - | 12 | 1523 | |||
BC017212.2 | - | 10 | 1225 | 4054134 | ||
BE621153.1 | - | 456 |