Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IRAK4 SIRNA Assays ›
Gene Symbol:
IRAK4
Gene Name:
interleukin 1 receptor associated kinase 4
Gene Aliases:
IPD1, IRAK-4, NY-REN-64, REN64
Chromosome Location:
Chr.12: 43758913 - 43789543 on Build GRCh38
UniGene ID:
Hs.138499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
134833
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IRAK4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114182.2
NP_001107654.1
10, 11
1253
NM_001145256.1
NP_001138728.1
9, 10
1107
NM_001145257.1
NP_001138729.1
8, 9
1059
NM_001145258.1
NP_001138730.1
7, 8
889
NM_016123.3
NP_057207.2
9, 10
1205
XM_005268943.3
XP_005269000.1
9, 10
1310
XM_005268944.4
XP_005269001.1
9, 10
1274
XM_005268945.4
XP_005269002.1
9, 10
1518
XM_005268947.4
XP_005269004.1
10, 11
1264
XM_005268948.2
XP_005269005.1
11, 12
1304
XM_005268949.2
XP_005269006.1
8, 9
1164
XM_006719438.3
XP_006719501.1
10, 11
1566
XM_006719439.2
XP_006719502.1
9, 10
1671
XM_006719440.2
XP_006719503.1
9, 10
1082
XM_006719441.2
XP_006719504.1
8, 9
967
XM_006719442.2
XP_006719505.1
9, 10
971
XM_006719443.3
XP_006719506.1
8, 9
1114
XM_011538431.2
XP_011536733.1
10, 11
1322
XM_011538432.1
XP_011536734.1
10, 11
1358
XM_011538433.2
XP_011536735.1
9, 10
1937
XM_017019390.1
XP_016874879.1
11, 12
1439
XM_017019391.1
XP_016874880.1
10, 11
1404
GenBank mRNA
AF155118.1
-
9, 10
1172
AF445802.1
-
8, 9
1123
AK000528.1
-
9, 10
1152
AK027301.2
-
9, 10
1700
AK299944.1
-
8, 9
1056
AK304583.1
-
7, 8
892
AK314288.1
-
9, 10
1194
AY283670.1
-
8, 9
1123
AY283671.1
-
8, 9
1121
AY340962.1
-
9, 10
1188
AY340963.1
-
10, 11
1236
AY340964.1
-
8, 9
1042
AY340965.1
-
9, 10
1090
AY340966.1
-
10, 11
1255
AY340967.1
-
11, 12
1303
AY730682.1
-
8, 9
1123
BC013316.1
-
9, 10
1193
4287014
KU178631.1
-
8, 9
1123
KU178632.1
-
7, 8
977
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
toll-like receptor signaling pathway
neutrophil mediated immunity
MyD88-dependent toll-like receptor signaling pathway
JNK cascade
cytokine-mediated signaling pathway
toll-like receptor 9 signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
positive regulation of smooth muscle cell proliferation
neutrophil migration
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
interleukin-1 receptor binding
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-9ljwx:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0