Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CPZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014447.2 | NP_001014447.1 | 10 | 1704 | ||
NM_001014448.2 | NP_001014448.1 | 10 | 1967 | |||
NM_003652.3 | NP_003643.2 | 9 | 1671 | |||
GenBank mRNA | AK056317.1 | - | 10 | 1703 | ||
AK127844.1 | - | 8 | 3147 | |||
BC006393.2 | - | 10 | 1871 | |||
BC018913.2 | - | 10 | 1871 | |||
BC080539.1 | - | 10 | 1638 | 5020194 | ||
U83411.1 | - | 9 | 1536 |