Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100594.1 | NP_001094064.1 | 6 | 3621 | ||
NM_017719.4 | NP_060189.3 | 7 | 3683 | |||
XM_005265245.3 | XP_005265302.1 | 8 | 3733 | |||
XM_011533861.2 | XP_011532163.1 | 5 | 2874 | |||
GenBank mRNA | AK025449.1 | - | 1 | 1808 | ||
AK026013.1 | - | 1 | 96 | |||
BC071567.1 | - | 6 | 3596 | 4375623 | ||
CR749621.1 | - | 7 | 3544 | |||
D43636.2 | - | 5 | 3354 |