Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016255.2 | NP_057339.1 | 5 | 1878 | ||
GenBank mRNA | AF097027.1 | - | 5 | 1810 | ||
AK074301.1 | - | 1 | 189 |