Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283035.1 | NP_001269964.1 | 3 | 347 | ||
NM_001283036.1 | NP_001269965.1 | 2 | 257 | |||
NM_001283037.1 | NP_001269966.1 | 2 | 257 | |||
NM_016407.4 | NP_057491.2 | 2 | 257 | |||
XM_017027872.1 | XP_016883361.1 | 5 | 450 | |||
GenBank mRNA | AF117231.1 | - | 2 | 205 | ||
AF161513.1 | - | 2 | 214 | |||
AF161518.1 | - | 2 | 183 | |||
AF212244.1 | - | 2 | 194 | |||
AK000481.1 | - | 2 | 187 | |||
AK000586.1 | - | 1 | 77 | |||
AK026118.1 | - | 2 | 185 | |||
AK225135.1 | - | 2 | 175 | |||
AK293662.1 | - | 2 | 185 | |||
AK302029.1 | - | 3 | 290 | |||
AK314563.1 | - | 2 | 212 | |||
BC003359.1 | - | 2 | 184 | 2900332 | ||
BG468509.1 | - | 2 | 218 | |||
BP307634.1 | - | 2 | 251 | |||
BU537939.1 | - | 2 | 167 | |||
CR983692.1 | - | 2 | 168 | |||
DC397043.1 | - | 3 | 347 |