Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278221.2 | NP_001265150.1 | 10 | 1807 | ||
NM_001321126.1 | NP_001308055.1 | 11 | 1883 | |||
NM_001321127.1 | NP_001308056.1 | 11 | 2016 | |||
NM_016483.6 | NP_057567.3 | 11 | 1924 | |||
GenBank mRNA | AF151060.2 | - | 9 | 972 | ||
AK223531.1 | - | 10 | 1141 | |||
AK292221.1 | - | 11 | 1730 | |||
AY014283.2 | - | 11 | 1498 | |||
BC022002.1 | - | 11 | 1783 | 4822315 |