Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMMDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016589.3 | NP_057673.2 | 5 | 745 | ||
GenBank mRNA | AF139077.1 | - | 5 | 718 | ||
AF210057.1 | - | 5 | 697 | |||
AL136622.1 | - | 5 | 720 | |||
AL390077.1 | - | 5 | 697 | |||
AL390090.1 | - | 5 | 672 | |||
AL390094.1 | - | 5 | 379 | |||
AY358633.1 | - | 5 | 698 | |||
BC012341.1 | - | 5 | 639 | 4576562 | ||
CR457158.1 | - | 5 | 532 | |||
CR533524.1 | - | 5 | 532 |