Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016608.1 | NP_057692.1 | 4 | 706 | ||
GenBank mRNA | AB039670.1 | - | 4 | 706 | ||
AF248963.1 | - | 4 | 639 | |||
AK074785.1 | - | 4 | 656 | |||
AK222578.1 | - | 4 | 683 | |||
BC002691.2 | - | 4 | 645 | 3605690 |