Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DACT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079520.1 | NP_001072988.1 | 4 | 847 | ||
NM_016651.5 | NP_057735.2 | 4 | 958 | |||
NR_046093.1 | 4 | 456 | ||||
NR_046095.1 | 4 | 764 | ||||
XM_006720167.3 | XP_006720230.1 | 4 | 895 | |||
GenBank mRNA | AF251079.2 | - | 1 | 63 | ||
AK299879.1 | - | 4 | 764 | |||
AK316164.1 | - | 4 | 554 | |||
AK316169.1 | - | 2 | 170 | |||
AK316237.1 | - | 4 | 456 | |||
AK316423.1 | - | 3 | 323 | |||
BX161433.1 | - | 4 | 411 | |||
DR157190.1 | - | 3 | 225 |