Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144893.1 | NP_001138365.1 | 5 | 762 | ||
NM_001144894.1 | NP_001138366.1 | 5 | 1038 | |||
NM_001144895.1 | NP_001138367.1 | 7 | 894 | |||
NM_001144896.1 | NP_001138368.1 | 6 | 1098 | |||
NM_001144897.1 | NP_001138369.1 | 7 | 1152 | |||
NM_001144899.1 | NP_001138371.1 | 7 | 687 | |||
NM_021155.3 | NP_066978.1 | 7 | 1170 | |||
NR_026692.1 | 6 | 1270 | ||||
XM_005272472.4 | XP_005272529.1 | 7 | 787 | |||
GenBank mRNA | AF290886.1 | - | 7 | 1112 | ||
AK293089.1 | - | 7 | 1126 | |||
AK304190.1 | - | 7 | 673 | |||
AK308595.1 | - | 4 | 713 | |||
AK313585.1 | - | 7 | 1171 | |||
AY042221.1 | - | 7 | 1103 | |||
AY042222.1 | - | 7 | 1085 | |||
AY042223.1 | - | 7 | 827 | |||
AY042224.1 | - | 6 | 395 | |||
AY042225.1 | - | 6 | 1031 | |||
AY042226.1 | - | 5 | 971 | |||
AY042227.1 | - | 6 | 695 | |||
AY042229.1 | - | 6 | 1203 | |||
AY042230.1 | - | 5 | 1131 | |||
AY042231.1 | - | 5 | 993 | |||
BC110615.2 | - | 5 | 1138 | 40032459 | ||
M98457.1 | - | 7 | 1144 |