Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144893.1 | NP_001138365.1 | 5 | 1064 | ||
NM_001144894.1 | NP_001138366.1 | 5 | 1340 | |||
NM_001144895.1 | NP_001138367.1 | 7 | 1196 | |||
NM_001144896.1 | NP_001138368.1 | 6 | 1400 | |||
NM_001144897.1 | NP_001138369.1 | 7 | 1454 | |||
NM_001144899.1 | NP_001138371.1 | 7 | 989 | |||
NM_021155.3 | NP_066978.1 | 7 | 1472 | |||
NR_026692.1 | 6 | 1572 | ||||
XM_005272472.4 | XP_005272529.1 | 7 | 1089 | |||
GenBank mRNA | AF290886.1 | - | 7 | 1414 | ||
AK293089.1 | - | 7 | 1428 | |||
AK304190.1 | - | 7 | 975 | |||
AK308595.1 | - | 4 | 1015 |