Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3KBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024666.2 | NP_001019837.1 | 4 | 424 | ||
NM_031892.2 | NP_114098.1 | 5 | 736 | |||
XM_005274494.2 | XP_005274551.1 | 5 | 602 | |||
XM_011545498.2 | XP_011543800.1 | 5 | 602 | |||
XM_011545499.2 | XP_011543801.1 | 4 | 410 | |||
XM_011545500.2 | XP_011543802.1 | 5 | 602 | |||
XM_011545503.2 | XP_011543805.1 | 5 | 602 | |||
XM_017029459.1 | XP_016884948.1 | 4 | 342 | |||
XM_017029460.1 | XP_016884949.1 | 4 | 385 | |||
XM_017029461.1 | XP_016884950.1 | 4 | 385 | |||
XM_017029462.1 | XP_016884951.1 | 5 | 602 | |||
XM_017029463.1 | XP_016884952.1 | 4 | 444 | |||
XM_017029464.1 | XP_016884953.1 | 5 | 677 | |||
XM_017029465.1 | XP_016884954.1 | 5 | 703 | |||
XM_017029466.1 | XP_016884955.1 | 5 | 602 | |||
XM_017029467.1 | XP_016884956.1 | 5 | 676 | |||
XM_017029468.1 | XP_016884957.1 | 5 | 703 | |||
GenBank mRNA | AF230904.1 | - | 5 | 711 | ||
AF542051.1 | - | 4 | 387 | |||
AK293234.1 | - | 4 | 364 | |||
AK313308.1 | - | 5 | 664 | |||
AY423734.1 | - | 6 | 634 | |||
AY542305.1 | - | 5 | 471 | |||
BC015806.1 | - | 5 | 638 | 3906722 | ||
BC050663.2 | - | 5 | 646 | |||
DC409010.1 | - | 4 | 424 |